missing translation for 'onlineSavingsMsg'
Learn More

Certified Reference Material Abel Flash point, Jet Aviation Fuel, UKAS ISO 17034 Certified 250mL (nominal value: 41.2 °C), Paragon Scientific™

Product Code. 16090040
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16090040 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16090040 Supplier Paragon Scientific Supplier No. CRMUABKR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Flash Point Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Abel Flash Point IP 170.

  • Fully certified in accordance with ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data currently available world-wide
  • Low levels of uncertainty achieves maximum accuracy of data
  • All data generated exclusively by UKAS ISO 17025 accredited laboratories
  • Fully traceable to international standards
  • Supplied in tamper evident UN compliant security pack
  • 12 month shelf life
  • Available from stock for immediate dispatch
  • Manufactured in the United Kingdom

Specifications

UN Number UN 1223
Quantity 250 mL
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 250ML CRMU-ABKR Abel Flash Point Standard, Jet Aviation Fuel
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use media other than water to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : kerosine

Nominal values quoted. Certified Values will vary from batch to batch.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.