Learn More
Certified Reference Material Cold Filter Plugging Point, Diesel, UKAS ISO 17034 Certified 250mL (Nominal value: -10.8 °C), Paragon Scientific™
Paragon CFPP Certified Reference Materials include products which are certified to and manufactured in strict accordance and compliance with ISO 17034
Brand: Paragon Scientific CRMU-CFGO1
Additional Details : CAS Number : 68334-30-5 Weight : 0.70000kg Transport : UN number : 1202 Chem class : 3 Pack group : III
Description
- Cold Filter Plugging Point (CFPP) Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Cold Filter Plugging Point (CFPP) according to IP 309. An alternative test method is ASTM D6371.
- Certified according to ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Specifications
UN 1202 | |
UKAS ISO 17034 |
Safety and Handling
- 250ML CRMU-CFGO1 Cold Filter Plugging Point Standard, Diesel
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Carcinogenicity Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : diesel fuel
Nominal values quoted. Certified Values will vary from batch to batch.