missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceruloplasmin Antibody (SR1152), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22625-100ul
This item is not returnable.
View return policy
Description
Ceruloplasmin Monoclonal antibody specifically detects Ceruloplasmin in Human, Rat samples. It is validated for Western Blot
Specifications
| Ceruloplasmin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| SR1152 | |
| Western Blot 1:500-1:2000 | |
| ceruloplasmin, ceruloplasmin (ferroxidase), CP-2, EC 1.16.3.1, Ferroxidase | |
| A synthesized peptide derived from human Ceruloplasmin (Uniprot #: P00450) | |
| 100 μg | |
| Cardiovascular Biology, Neurodegeneration, Neuroscience | |
| 1356 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction