missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceruloplasmin Antibody (SR1152), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22625-50ul
This item is not returnable.
View return policy
Description
Ceruloplasmin Monoclonal antibody specifically detects Ceruloplasmin in Human, Rat samples. It is validated for Western Blot
Specifications
| Ceruloplasmin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| SR1152 | |
| Western Blot 1:500-1:2000 | |
| ceruloplasmin, ceruloplasmin (ferroxidase), CP-2, EC 1.16.3.1, Ferroxidase | |
| A synthesized peptide derived from human Ceruloplasmin (Uniprot #: P00450) | |
| 50 μg | |
| Cardiovascular Biology, Neurodegeneration, Neuroscience | |
| 1356 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur