missing translation for 'onlineSavingsMsg'
Learn More

Cetalkonium Chloride, TRC

Product Code. 30004763
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004763 1 g 1g
30134957 10 g 10g
17994715 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004763 Supplier Toronto Research Chemicals Supplier No. TRCC2805001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-18-9
Formula Weight 395.3319
InChI Formula InChI=1S/C25H46N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-20-23-26(2,3)24-25-21-18-17-19-22-25;/h17-19,21-22H,4-16,20,23-24H2,1-3H3;1H/q+1;/p-1
IUPAC Name benzyl-hexadecyl-dimethylazanium;chloride
Molecular Formula C25 H46 N . Cl
Molecular Weight (g/mol) 396.09
Percent Purity >95%
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1
Synonym Ammonium, benzylhexadecyldimethyl-, chloride (8CI), Benzenemethanaminium, N-hexadecyl-N,N-dimethyl-, chloride (9CI), Benzylhexadecyldimethylammonium chloride (6CI), Acetoquate CDAC, Acinol, Acquat CDAC, Ammonyx G, Ammonyx T, Baktonium, Banicol, Benzaletas, Benzyl-n-hexadecyldimethylammonium chloride, Benzylcetyldimethylammonium chloride, Benzyldimethylcetylammonium chloride, Benzyldimethylhexadecylammonium chloride, Bicetonium, Bonjela, CDBAC, Cetalkonium chloride, Cetol, Cetyl zephiran, Cetylbenzyldimethylammonium chloride, Cetyldimethylbenzylammonium chloride, Cetylon, DMCBAC, Dehyquart CBB, Dehyquart CDB, Dimethylbenzylcetylammonium chloride, Dimethylbenzylhexadecylammonium chloride, HBAC, Hexadecylbenzyldimethylammonium chloride, Hexadecyldimethylbenzylammonium chloride, N-Benzyl-N-cetyldimethylammonium chloride, N-Hexadecyl-N,N-dimethyl-N-benzylammonium chloride, N-Hexadecyl-N-benzyl-N,N-dimethylammonium chloride, Pharycidin Concentrate, Querton 16BCL, Sanisol C, Spilan, Tetraseptan, Win 357, Zettyn, Zettyn chloride, n-Hexadecyldimethylbenzylammonium chloride
Chemical Name or Material Benzyldimethylhexadecylammonium Chloride
Grade HPLC
Show More Show Less

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.