Learn More
Cetuximab Mouse, Biotin, UltraLys, Monoclonal Antibody, AcroBiosystems
Mouse Monoclonal Antibody
Brand:  ACROBiosystems CEB-BY31-25ug
 View more versions of this product
Product Code. 16447409
  • 414.00 € / 25µg
Description
Description
Cetuximab Monoclonal antibody specifically detects Cetuximab, recommended for PK/PD.
Specifications
Specifications
Cetuximab
Biotin
25 μg
Primary
Monoclonal
Hybridoma
RUO
-20°C