missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFAP299 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
386.00 € - 559.00 €
Specifications
| Antigen | C4orf22 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18444921
|
Novus Biologicals
NBP1-86203-25ul |
25 μL |
386.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18205178
|
Novus Biologicals
NBP1-86203 |
0.1 mL |
559.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CFAP299 Polyclonal specifically detects CFAP299 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| C4orf22 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| chromosome 4 open reading frame 22, hypothetical protein LOC255119, MGC35043 | |
| CFAP299 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 255119 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QEEGLKALDNIVTQFNAYEDFLDSQITTVDLYYLEDETLARQLVELGYRGTGERVKREDFEARKAAIEIARLAERAQQKTLTSAGK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title