missing translation for 'onlineSavingsMsg'
Learn More

CFTR Mouse, Clone: CFTR/1342, enQuireBio™

Mouse Monoclonal Antibody

199.00 € - 478.00 €

Specifications

Antigen CFTR
Clone CFTR/1342
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15989747
View Documents
EnquireBio
1080-MSM2-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15999747
View Documents
EnquireBio
1080-MSM2-P1
100 μg
ONLINE EXCLUSIVE PRICE
554.40 €
478.00 € / 100µg
Save 76.40 €
13% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CFTR
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% Azide
1080
Recombinant human full length CFTR
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
165-170kDa
CFTR/1342
Immunohistochemistry
Unconjugated
RUO
NP_011514056
CFTR
IgG2a
Protein A/G Purified
Cell Surface and Cytoplasmic
MOLT-4 cells. Pancreas, Kidney or Placenta.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.