missing translation for 'onlineSavingsMsg'
Learn More

CFTR Mouse, Clone: SPM176, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 1080-MSM2X-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929757

  • 485.00 € / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CFTR
Cell Surface and Cytoplasmic
SPM176
Unconjugated
NP_011514056
Mouse
165-170kDa
Protein A/G Purified
RUO
1080
IgG2a
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
CFTR
Recombinant human full-length CFTR
MOLT-4 cells. Pancreas, Kidney or Placenta.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers