missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CGBP Recombinant Rabbit Monoclonal Antibody (K01_1P50)

Product Code. 30830401
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830401 50 μL
30828445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830401 Supplier Invitrogen™ Supplier No. MA564958

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CGBP Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

Proteins that contain a CXXC motif within their DNA-binding domain, such as CXXC1, recognize CpG sequences and regulate gene expression (Carlone and Skalnik, 2001).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGBP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1P50
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene CXXC1
Gene Accession No. Q9CWW7, Q9P0U4
Gene Alias 2410002I16Rik; 5830420C16Rik; AI426635; CFP 1; CFP1; CGBP; CpG binding protein; cpG-binding protein; cpG-binding protein; CXXC-type zinc finger protein 1; CXXC 1; CXXC finger 1; CXXC finger 1 (PHD domain); CXXC finger protein 1; Cxxc1; CXXC-type zinc finger protein 1; DNA-binding protein with PHD finger and CXXC domain; hCGBP; HsT2645; PCCX 1; Pccx1; PHD finger and CXXC domain-containing protein 1; PHF 18; PHF18; SPP 1; SPP1; ZCGPC 1; ZCGPC1; zinc finger, CpG binding-type containing 1
Gene Symbols CXXC1
Host Species Rabbit
Immunogen A synthetic peptide of CGBP.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291440, 30827, 74322
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.