missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGI-09 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 463.00 €
Specifications
| Antigen | CGI-09 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18685052
|
Novus Biologicals
NBP2-92578-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683362
|
Novus Biologicals
NBP2-92578-0.1ml |
0.1 mL |
463.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CGI-09 Polyclonal antibody specifically detects CGI-09 in Human samples. It is validated for Western BlotSpecifications
| CGI-09 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CGI-09, GCD10, Gcd10p, KIAA1153, MGC5029, TRM6, tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6, tRNA methyltransferase 6 homolog (S. cerevisiae), tRNA(m1A58)-methyltransferase subunit TRM6, tRNA(m1A58)MTase subunit TRM6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human TRMT6 (NP_057023.2). MEGSGEQPGPQPQHPGDHRIRDGDFVVLKREDVFKAVQVQRRKKVTFEKQWFYLDNVIGHSYGTAFEVTSGGSLQPKKKREEPTAETKEAGTDNRNIVDDGKSQKLTQDDIKALKDKGIKGEEIVQQLIENSTTFRDKTEFAQDKYIKKKKKKYEAIITVVKPSTRILSIMYYAREPGKINHMRYDTLAQMLTLGNIRAG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 51605 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title