Learn More
ChABC Antibody (1E10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NBP1-96141SS
 View more versions of this product
Product Code. 18759723
  • 226.00 € / 25µL
Description
ChABC Monoclonal specifically detects ChABC in Rat, Non-species specific samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
80 kDa
Protein G purified
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.