missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHCHD3 Monoclonal Antibody (24GB6560)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557361
This item is not returnable.
View return policy
Description
CHCHD3 Monoclonal Antibody for Western Blot
Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintenance of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Has also been shown to function as a transcription factor which binds to the BAG1 promoter and represses BAG1 transcription. Plays an important role in the maintenance of the MICOS complex stability and the mitochondrial cristae morphology.
Specifications
| CHCHD3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9CRB9, Q9NX63 | |
| CHCHD3 | |
| Recombinant protein of human CHCHD3 (aa 2-227). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| 24GB6560 | |
| Unconjugated | |
| CHCHD3 | |
| 0610041L09Rik; 1700039J09Rik; AW558177; CHCHD3; coiled-coil-helix-coiled-coil-helix domain containing 3; Coiled-coil-helix-coiled-coil-helix domain-containing protein 3; coiled-coil-helix-coiled-coil-helix domain-containing protein 3, mitochondrial; FLJ20420; Mic19; MICOS complex subunit MIC19; MINOS3; mitochondrial inner membrane organizing system 3; PPP1R22; protein phosphatase 1, regulatory subunit 22 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 296966, 54927, 66075 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction