missing translation for 'onlineSavingsMsg'
Learn More

Chebulinic acid, MedChemExpress

Product Code. 16461668
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461668 5 mg 5mg
16409258 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461668 Supplier Medchem Express Supplier No. HYN2033
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chebulinic acid is a potent natural inhibitor of M. tuberculosis DNA gyrase, also can inhibit SMAD-3 phosphorylation, inhibit H+ K+-ATPase activity.

Chebulinic acid is a potent natural inhibitor of M. tuberculosis DNA gyrase, also can inhibit SMAD-3 phosphorylation, inhibit H+ K+-ATPase activity.

Specifications

Chemical Name or Material Chebulinic acid
CAS 18942-26-2
Color White
Purity Grade Notes Research
Molecular Formula C41H32O27
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (104.53 mM; ultrasonic and warming and heat to 60°C)
SMILES OC(C[C@H](C(O[C@@]([C@H](O[C@H]1OC(C2=CC(O)=C(O)C(O)=C2)=O)COC(C3=CC(O)=C(O)C(O)=C3)=O)([H])[C@@](OC(C4=CC(O)=C(O)C(O)=C4)=O)([H])[C@@]1([H])OC5=O)=O)[C@@]([C@H]6O)([H])C(C5=CC(O)=C7O)=C7OC6=O)=O
Molecular Weight (g/mol) 956.68
Formula Weight 956.68
Percent Purity 97.01%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.