missing translation for 'onlineSavingsMsg'
Learn More

Chebulinic Acid, TRC

Product Code. 30009772
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009772 5 mg 5mg
30005901 1 mg 1mg
30011177 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009772 Supplier Toronto Research Chemicals Supplier No. TRCC2918955MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18942-26-2
Molecular Formula C41 H32 O27
Molecular Weight (g/mol) 956.68
Synonym β-D-Glucopyranose, 1,3,6-tris(3,4,5-trihydroxybenzoate), cyclic 2→2:4→1-ester with (2S)-[(3R,4S)-5-carboxy-3,4-dihydro-3,7,8-trihydroxy-2-oxo-2H-1-benzopyran-4-yl]butanedioic acid, 7,11-Methanopyrano[4,3,2-kl][2,5,8]benzotrioxacyclotridecin, β-D-glucopyranose deriv., Chebulinic acid, Eutannin, NSC 69862
IUPAC Name 2-[(4R,5S,7R,8R,11S,12S,13S,21S)-13,17,18-trihydroxy-2,10,14-trioxo-5,21-bis[(3,4,5-trihydroxybenzoyl)oxy]-7-[(3,4,5-trihydroxybenzoyl)oxymethyl]-3,6,9,15-tetraoxatetracyclo[10.7.1.14,8.016,20]henicosa-1(19),16(20),17-trien-11-yl]acetic acid
SMILES O[C@@H]1[C@H]2[C@H](CC(=O)O)C(=O)O[C@@H]3[C@@H](COC(=O)c4cc(O)c(O)c(O)c4)O[C@@H](OC(=O)c5cc(O)c(O)c(O)c5)[C@H](OC(=O)c6cc(O)c(O)c(OC1=O)c26)[C@H]3OC(=O)c7cc(O)c(O)c(O)c7

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C41H32O27/c42-15-1-10(2-16(43)26(15)51)35(56)62-9-22-31-33(66-36(57)11-3-17(44)27(52)18(45)4-11)34(41(63-22)68-37(58)12-5-19(46)28(53)20(47)6-12)67-38(59)13-7-21(48)29(54)32-25(13)24(30(55)40(61)65-32)14(8-23(49)50)39(60)64-31/h1-7,14,22,24,30-31,33-34,41-48,51-55H,8-9H2,(H,49,50)/t14-,22+,24-,30+,31+,33-,34+,41-/m0/s1
Formula Weight 956.1131
Chemical Name or Material Chebulinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.