Learn More
Check Standards, Palintest™
Product Code. 13188780
Change view
Click to view available options
For Use With (Equipment):
Ammonia Meter 1000
Bromometer Duo
Chlordiox-Duo
Chloridiox Meter 1000
Ozone Meter 1000
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13188780 Chloridiox Meter 1000 Each
11342900 Ammonia Meter 1000 Each
13168780 Bromometer Duo -
13178780 Chlordiox-Duo Each
10119459 Ozone Meter 1000 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13188780 Supplier Palintest Supplier No. PT 229/M2
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Check the performance of your Palintest™; instument with specific standards. Palintest™; Check Standards are traceable standards that ensure testing accuracy.
  • Traceable reference materials defined in terms of % transmittance (%T) at a particular wavelength.
  • Ideal in confirming that a Palintest™ photometer is conforming to specification.
  • Each standard enables users to check each of the individual wavelengths in the photometer.
Specifications
Description Check Standards for Chloridiox Meter 1000
For Use With (Equipment) Chloridiox Meter 1000
Type Check Standard