missing translation for 'onlineSavingsMsg'
Learn More

CHES, 0.5M buffer soln, pH 10.0, Thermo Scientific™

Product Code. 15415669
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
Product Code. Quantity unitSize
15415669 100 mL 100mL
15425669 250 mL 250mL
2 options
This item is not returnable. View return policy

Product Code. 15415669

Brand: Thermo Scientific Alfa Aesar J63419.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-47-9
Molecular Formula C8H17NO3S
Molecular Weight (g/mol) 207.288
MDL Number MFCD00003835
InChI Key MKWKNSIESPFAQN-UHFFFAOYSA-N
PubChem CID 66898
IUPAC Name 2-(cyclohexylamino)ethanesulfonic acid
SMILES C1CCC(CC1)NCCS(=O)(=O)O

Specifications

Concentration 0.5 M
pH 10.0
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material CHES buffer soln.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.