missing translation for 'onlineSavingsMsg'
Learn More

Melford CHES

Product Code. 30018151
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
30018151 100 g 100g
30018152 500 g 500g
2 options
This item is not returnable. View return policy

Product Code. 30018151

Brand: Melford C40020100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHES, also known as 2-(N-Cyclohexylamino) ethanesulfonic acid, is a chemical compound that plays a significant role in life science research, particularly in molecular biology and protein research.

CHES is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures. It is particularly effective in the pH range of 8.6 to 10.0, making it suitable for applications requiring higher alkaline pH conditions.

CHES is also commonly used in protein purification, enzyme assays, nucleic acid research, and electrophoresis techniques where a specific pH range needs to be maintained. Its buffering capacity and stability in alkaline conditions make it valuable in these applications.

One of the significant advantages of CHES is its low ionic strength, which minimizes interference with biological processes and enzymatic activities. It allows for accurate and reliable results without compromising the integrity of the biological samples or the efficiency of the enzymatic reactions.

Specifications

Chemical Name or Material CHES
CAS 103-47-9
Quantity 100 g
Molecular Formula C8H17NO3S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.