missing translation for 'onlineSavingsMsg'
Learn More

Chicken IgY Mouse anti-Chicken, HRP, Clone: 1E12G2, Proteintech

Product Code. 17581786 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17581786 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17581786 Supplier Proteintech Supplier No. HRP67405100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The Chicken IgY antibody from Proteintech is a mouse monoclonal antibody to a peptide of chicken Chicken IgY. This antibody recognizes chicken antigen. The Chicken IgY antibody has been validated for the following applications: analysis. This antibody is hrp.

Specifications

Antigen Chicken IgY
Classification Monoclonal
Clone 1E12G2
Concentration 2000 μg/mL
Conjugate HRP
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Target Species Chicken
Isotype IgG2a
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.