missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Chicken Universal Protein Whole Tissue Lysate (Adult Whole Normal)

Product Code. 18236593 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18236593 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18236593 Supplier Novus Biologicals™ Supplier No. NBL126502
  • ONLINE EXCLUSIVE PRICE
    690.00 €
    652.05 € / 0.5mg
    Save 37.95 €
    5% Off
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The Chicken Universal Protein Whole Tissue Lysate (Adult Whole Normal) is derived from chicken.
TRUSTED_SUSTAINABILITY

Specifications

Tissue Universal
Quantity 0.5 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Universal Protein Lysate: Chicken Normal Tissues
Cellular Fraction Whole
Tissue Condition Normal
Cross Reactivity Chicken
Host Species Chicken
Product Type Chicken Universal Protein Whole Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.