missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHID1 Antibody (CHID1/6368), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23888-20ug
This item is not returnable.
View return policy
Description
CHID1 Monoclonal antibody specifically detects CHID1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CHID1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| chitinase domain containing 1, chitinase domain-containing protein 1, FLJ42707, MGC3234, SI-CLP, stabilin-1 interacting chitinase-like protein, Stabilin-1-interacting chitinase-like protein | |
| Recombinant full-length human CHID1 protein | |
| 20 μg | |
| Lipid and Metabolism | |
| 66005 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| CHID1/6368 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction