missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHIL3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547323
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The CHIL3 gene encodes the protein chitinase-like 3, which is primarily secreted by macrophages and is involved in immune regulation, tissue remodeling, and inflammatory responses. CHIL3, also known as Ym1, belongs to the chitinase-like protein family but lacks chitinase activity due to mutations in its catalytic domain. It is predominantly expressed in lung tissue, where it plays a vital role in asthmatic responses and allergic inflammation. Furthermore, CHIL3 exerts anti-inflammatory roles during pathogen infection by modulating macrophage pro-inflammatory activity and promoting the Th2 immune response. Beyond immunological involvement, CHIL3 is integral to tissue repair mechanisms and cancer cell proliferation, contributing to extracellular matrix restructuring and angiogenesis. It facilitates macrophage polarization towards an M2 phenotype, which is associated with tumor progression and metastasis. Thus, CHIL3 is considered a potential biomarker for inflammatory diseases and cancer, with ongoing research examining its therapeutic modulation.
Specifications
| CHIL3 | |
| Polyclonal | |
| Unconjugated | |
| Chil3 | |
| AI505981; beta-N-acetylhexosaminidase Ym1; Chi3l3; Chil3; chitinase 3-like 3; chitinase-3-like protein 3; chitinase-like 3; chitinase-like protein 3; ECF-L; eosinophil chemotactic cytokine; eosinophil chemotactic factor-L; secreted protein Ym1; Ym1 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 12655 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O35744 | |
| Chil3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Chitinase 3-like 3/ECF-L Tyr22-Tyr398. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction