Learn More
Chinese Hamster PLBD2 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB106921-100
 View more versions of this product
Product Code. 18122271
  • 517.00 € / 100µg
Description
PLBD2 Monoclonal antibody specifically detects PLBD2 in Chinese Hamster samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PLBD2
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
LAMA-like protein 2,76 kDa protein, lamina ancestor homolog 2, mannose-6-phosphate protein associated protein p76, P76, phospholipase B domain containing 2, phospholipase B domain-containing protein 2, phospholipase B-like 2 32 kDa form, phospholipase B-like 2 45 kDa form, PLB homolog 2, putative phospholipase B-like 2
Recombinant chinese hamster PLBD-2, Met1-Asp585, Accession # XP_003510860
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG
ELISA
2751B
ELISA
XP_003510860
Rabbit
Protein A or G purified from hybridoma culture supernatant
RUO
196463
Chinese Hamster
Purified