missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CHIR 99021 trihydrochloride
GREENER_CHOICE

Product Code. 18769007
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18769007

Brand: Tocris Bioscience™ 4953/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrochloride salt of CHIR 99021 (Cat. No. 4423); selective GSK-3 inhibitor

Hydrochloride salt of CHIR 99021 (Cat. No. 4423), a highly selective GSK-3 inhibitor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1782235-14-6
Molecular Formula C22H21Cl5N8

Specifications

Quantity 50 mg
Formula Weight 574.72
Purity >99%
Molecular Weight (g/mol) 574.72
Chemical Name or Material CHIR 99021 trihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.