Learn More
Chitinase 3-like 1/YKL-40 Antibody (CL12241) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21212IR
Product Code. 30287587
  • 737.00 € / 0.10mL
Description
Chitinase 3-like 1/YKL-40 Monoclonal antibody specifically detects Chitinase 3-like 1/YKL-40 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chitinase 3-like 1/YKL-40
Monoclonal
DyLight 755
AW208766, Brp39, Chi3l1, gp39, YKL-40
This antibody was developed using a synthetic peptide derived from P36222, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cytoskeleton Markers, Signal Transduction
1116
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL12241
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified