missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chitinase 3-like 2 Antibody (1B2.G4), Novus Biologicals™
Mouse Monoclonal Antibody
256.00 € - 484.00 €
Specifications
| Antigen | Chitinase 3-like 2 |
|---|---|
| Clone | 1B2.G4 |
| Concentration | 0.2 mg/ml |
| Applications | ELISA, Functional Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30302898
|
Novus Biologicals
NBP3-44589-20ug |
20 μg |
256.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30294631
|
Novus Biologicals
NBP3-44589-100ug |
100 μg |
484.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chitinase 3-like 2 Monoclonal antibody specifically detects Chitinase 3-like 2 in Human samples. It is validated for ELISA,Functional AssaySpecifications
| Chitinase 3-like 2 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| chitinase 3-like 2, chitinase-3-like protein 2, Chondrocyte protein 39, YKL-39YKL39 | |
| Recombinant human fragment corresponding to Chitinase 3-like 2 protein (Uniprot: Q15782). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| 1B2.G4 | |
| ELISA, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Stem Cells | |
| 10 mM PBS with 0.05% BSA | |
| 1117 | |
| IgG2 κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title