missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chk1, p Ser296 Antibody (SR1961), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21549-100ul
This item is not returnable.
View return policy
Description
Chk1 Monoclonal antibody specifically detects Chk1 in Human samples. It is validated for Western Blot
Specifications
| Chk1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1961 | |
| Western Blot 1:500-1:2000 | |
| Checkpoint, S. pombe, homolog of, 1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1 | |
| A synthesized peptide derived from human phospho-Chk1 [Ser296] (Uniprot #: O14757) | |
| 100 μg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers, Tumor Suppressors | |
| 1111 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction