missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloramphenicol Acetyltransferase Antibody (CAT-1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50129-0.025ml
This item is not returnable.
View return policy
Description
Chloramphenicol Acetyltransferase Monoclonal antibody specifically detects Chloramphenicol Acetyltransferase in Nonspecies specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Chloramphenicol Acetyltransferase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| CAT, CAT II, CATB, CatD, Chloramphenicol acetyltransferase 2, Chloramphenicol acetyltransferase II, cmlA | |
| Full length Chloramphenicol Acetyltransferase | |
| 0.025 mL | |
| Epitope Tags | |
| Non-species specific | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| CAT-1 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction