missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol (Crystalline Powder), Fisher BioReagents

Product Code. 10255203
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10255203 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10255203 Supplier Fisher BioReagents Supplier No. BP904100
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For selecting transformed cells containing chloramphenicol resistance genes

Used as a selection agent for transformed cells containing chloramphenicol resistance genes. Mode of Action: Inhibits translation on the 50S ribosomal subunit at the peptidyltransferase step (elongation inhibition).
TRUSTED_SUSTAINABILITY

Specifications

Type Chloramphenicol
Physical Form Solid
CAS 56-75-7
pH 5 to 7
Melting Point 149°C
Packaging Poly Bottle
Specific Rotation Inclusive between 17° (+ or -)
Color White
Quantity 100 g
Assay Percent Range 97%
Molecular Formula C11H12Cl2N2O5
Merck Index 15, 2077
Synonym Chloromycetin, D-(?)-threo-2, 2-Dichloro-N-[?-hydroxy-?-(hydroxymethyl)-?-(4-nitrophenyl)ethyl]acetamide
InChI Key WIIZWVCIJKGZOK-RKDXNWHRSA-N
SMILES C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
Molecular Weight (g/mol) 323.126
PubChem CID 5959
ChEBI CHEBI:17698
Formula Weight 323.13
Chemical Name or Material Chloramphenicol
Show More Show Less
compliance-icons
Product Identifier
  • Chloramphenicol
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.