missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol, Extra Pure, SLR, Fisher Chemical

Product Code. 10324980 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10324980 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10324980 Supplier Fisher Chemical Supplier No. C/4322/47
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Physical Form Solid
CAS 56-75-7
pH 5 to 7
Melting Point 149°C
Packaging Plastic Powder Jar
Color White
Quantity 50 g
Molecular Formula C11H12Cl2N2O5
MDL Number 7360
Formula Weight 323.13g/mol
compliance-icons
Product Identifier
  • Chloramphenicol
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.