missing translation for 'onlineSavingsMsg'
Learn More

Chloranilic acid, 99+%, Thermo Scientific™

Product Code. 10204280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10204280 100 g Glass bottle 100g
10677484 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10204280 Supplier Thermo Scientific Acros Supplier No. 108481000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-88-7
MDL Number MFCD00001596
InChI Key IPPWILKGXFOXHO-UHFFFAOYSA-N
Synonym chloranilic acid, 2,5-dichloro-3,6-dihydroxy-p-benzoquinone, p-chloranilic acid, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy, unii-yj8l3bb7y4, 2,5-dichloro-3,6-dihydroxybenzoquinone, 2,5-dihydroxy-3,6-dichlorobenzoquinone, yj8l3bb7y4, p-quinone, 2,5-dichloro-3,6-dihydroxy, p-benzoquinone, 2,5-dichloro-3,6-dihydroxy
PubChem CID 66604
IUPAC Name 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione
SMILES C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O

Specifications

Melting Point 282°C to 284°C
Color Orange to Red
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C6H2Cl2O4
Quantity 100 g
Beilstein 08, 379
Merck Index 15, 2079
Solubility Information Solubility in water: soluble in hot water
Molecular Weight (g/mol) 208.99
Formula Weight 208.99
Percent Purity 99+%
Physical Form Fine Powder
Chemical Name or Material Chloranilic acid
Show More Show Less
compliance-icons
Product Identifier
  • Chloranilic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.