missing translation for 'onlineSavingsMsg'
Learn More

Chlordane (Mixture of Isomers) 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30312513
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312513 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312513 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010331ME
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlordane
CAS 57-74-9
Analyte or Component Names Chlordane
Shipping Condition Room Temperature
Molecular Formula C10 H6 Cl8
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Chlordane, Chlordan, 1,2,4,5,6,7,8,8-Octachloro-2,3,3a,4,7,7a-hexahydro-4,7-methano-1H-indene, 1,2,4,5,6,7,8,8-Octachloro-3a,4,7,7a-tetrahydro-4,7-methanoindan, 1,2,4,5,6,7,10,10-Octachloro-4,7,8,9-tetrahydro-4,7-endomethyleneindane, 1,2,4,5,6,7,10,10-Octachloro-4,7,8,9-tetrahydro-4,7-methyleneindane, 1,2,4,5,6,7,8,8-Octachloro-4,7-methano-3a,4,7,7a-tetrahydroindane, CD 68, Chlorindan, Cortilan-neu, Dowchlor, ENT 9932, HCS 3260, M 140, NSC 8931, Octachloro-4,7-methanotetrahydroindane, Oktaterr, Tat Chlor 4, Toxichlor
InChI Formula InChI=1S/C10H6Cl8/c11-3-1-2-4(5(3)12)9(16)7(14)6(13)8(2,15)10(9,17)18/h2-5H,1H2/t2?,3?,4?,5?,8-,9+/m0/s1
SMILES ClC1CC2C(C1Cl)[C@@]3(Cl)C(=C(Cl)[C@]2(Cl)C3(Cl)Cl)Cl
IUPAC Name 1,2,4,5,6,7,8,8-Octachloro-2,3,3a,4,7,7a-hexahydro-4,7-methano-1H-indene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 409.78
Quantity 1 mL
Formula Weight 405.7978 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.