missing translation for 'onlineSavingsMsg'
Learn More

Chlordane (Mixture of isomers), TRC

Product Code. 30138134
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138134 25 mg 25mg
17987639 5 mg 5mg
17999439 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138134 Supplier Toronto Research Chemicals Supplier No. TRCC32703025MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-74-9
Molecular Formula C10 H6 Cl8
Molecular Weight (g/mol) 409.78
Synonym Chlordane, Chlordan, 1,2,4,5,6,7,8,8-Octachloro-2,3,3a,4,7,7a-hexahydro-4,7-methano-1H-indene, 1,2,4,5,6,7,8,8-Octachloro-3a,4,7,7a-tetrahydro-4,7-methanoindan, 1,2,4,5,6,7,10,10-Octachloro-4,7,8,9-tetrahydro-4,7-endomethyleneindane, 1,2,4,5,6,7,10,10-Octachloro-4,7,8,9-tetrahydro-4,7-methyleneindane, 1,2,4,5,6,7,8,8-Octachloro-4,7-methano-3a,4,7,7a-tetrahydroindane, CD 68, Chlorindan, Cortilan-neu, Dowchlor, ENT 9932, HCS 3260, M 140, NSC 8931, Octachloro-4,7-methanotetrahydroindane, Oktaterr, Tat Chlor 4, Toxichlor
IUPAC Name 1,2,4,5,6,7,8,8-Octachloro-2,3,3a,4,7,7a-hexahydro-4,7-methano-1H-indene
SMILES ClC1CC2C(C1Cl)[C@@]3(Cl)C(=C(Cl)[C@]2(Cl)C3(Cl)Cl)Cl

Specifications

Analyte or Component Names Chlordane
Shipping Condition Room Temperature
InChI Formula InChI=1S/C10H6Cl8/c11-3-1-2-4(5(3)12)9(16)7(14)6(13)8(2,15)10(9,17)18/h2-5H,1H2/t2?,3?,4?,5?,8-,9+/m0/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 25 mg
Formula Weight 405.7978
Chemical Name or Material Chlordane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.