missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine (digluconate), MedChemExpress

Product Code. 16421028
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16421028 5 g 5g
16458386 100 mg 100mg
16412249 1 g 1g
16402186 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16421028 Supplier Medchem Express Supplier No. HYB0608
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chlorhexidine digluconate is an antiseptic effective against a wide variety of gram-negative and gram-positive organisms.

Chlorhexidine digluconate is an antiseptic effective against a wide variety of gram-negative and gram-positive organisms.

Specifications

Chemical Name or Material Chlorhexidine (digluconate)
Physical Form Liquid
CAS 18472-51-0
Color Colorless
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C34H54Cl2N10O14
For Use With (Application) COVID-19-anti-virus
Solubility Information H2O : 100 mg/mL (111.39 mM; Need ultrasonic) ∣DMSO : ≥ 38 mg/mL (42.33 mM)
SMILES N=C(NC(NCCCCCCNC(NC(NC1=CC=C(Cl)C=C1)=N)=N)=N)NC2=CC=C(Cl)C=C2.O[C@H]([C@@H](O)C(O)=O)[C@H](O)[C@H](O)CO.O[C@H]([C@@H](O)C(O)=O)[C@H](O)[C@H](O)CO
Molecular Weight (g/mol) 897.76
Formula Weight 897.76
Percent Purity 98.15%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.