missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine, MedChemExpress

Product Code. 16438706
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438706 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438706 Supplier Medchem Express Supplier No. HYB1248100mg
 Restricted Item
Estimated Shipment: 02-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chlorhexidine is an antibacterial used as an antiseptic and for other applications. Chlorhexidine is used to clean the skin after an injury, before surgery, or before an injection. Chlorhexidine is also used to clean the hands before a procedure.

Chlorhexidine is an antibacterial used as an antiseptic and for other applications. Chlorhexidine is used to clean the skin after an injury, before surgery, or before an injection. Chlorhexidine is also used to clean the hands before a procedure.

Specifications

Chemical Name or Material Chlorhexidine
Physical Form Solid
CAS 55-56-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H30Cl2N10
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 25 mg/mL (49.46 mM; Need ultrasonic)
SMILES N=C(NC1=CC=C(Cl)C=C1)NC(NCCCCCCNC(NC(NC2=CC=C(Cl)C=C2)=N)=N)=N
Molecular Weight (g/mol) 505.45
Formula Weight 505.45
Percent Purity 99.46%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.