missing translation for 'onlineSavingsMsg'
Learn More

Chloride Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Brand:  Fisher Chemical A 013 500

Code : 04

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 12982105

  • 107.00 € / 500mL
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1g/L
500 mL
H2O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers