missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloride Standard Solution 1000ppm (1mg/mL), for Ion Pair Chromatography, ready to use, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Packaging | HDPE plastic bottle |
| CAS | 7447-40-7 |
| Physical Form | Liquid |
| Molecular Formula | ClK |
| MDL Number | MFCD00011360 |
| Color | Colorless |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| SMILES | [Cl-].[K+] |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction