Learn More
Chlorinated Hydrocarbons Mix, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep 612-X
Code : NEW
Additional Details : Weight : 0.00100kg Transport : UN number : 1262 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
1,2,4-Trichlorobenzene | 120-82-1 | 2000μg/mL |
1,2-Dichlorobenzene | 95-50-1 | 2000μg/mL |
1,3-Dichlorobenzene | 541-73-1 | 2000μg/mL |
1,4-Dichlorobenzene | 106-46-7 | 2000μg/mL |
2-Chloronaphthalene | 91-58-7 | 2000μg/mL |
Hexachlorobenzene | 118-74-1 | 2000μg/mL |
Hexachlorobutadiene | 87-68-3 | 2000μg/mL |
Hexachlorocyclopentadiene | 77-47-4 | 2000μg/mL |
Hexachloroethane | 67-72-1 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Chlorinated Hydrocarbons Mix | |
2mL amber prescored glass ampule | |
Isooctane | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
USEPA Method 612, Method 8120/8120A | |
For USEPA Method 612, 8120, 8120A, 8121 | |
1 mL | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
Safety and Handling
- Chlorinated Hydrocarbons Mix
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : hexachlorobuta-1,3-diene, 1,2,4-trichlorobenzene
Product intended for laboratory use only