missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorinated Organic and Desinfectant by-products Mix 1 100 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| CAS | 71-55-6, 513-88-2, 96-12-8, 106-93-4, 83463-62-1, 75-27-4 |
| Quantity | 1 mL |
| Concentration | Dibromoacetonitrile 99.8 μg/mL∣Bromochloroacetonitrile 99.9 μg/mL∣1,2-Dibromo-3-chloropropane 100 μg/mL∣Chloropicrin 100 μg/mL∣Trichloroethene 100 μg/mL∣1,1-Dichloropropanone-2 100.1 μg/mL∣1,2-Dibromoethane 100.1 μg/mL∣Bromodichloromethane 100.1 μg/mL∣Chloroform 100.1 μg/mL∣Dibromochloromethane 100.1 μg/mL∣Tetrachloroethene 100.1 μg/mL∣Tetrachloromethane 100.1 μg/mL∣Tribromomethane 100.1 μg/mL∣Trichloroacetonitrile 100.1 μg/mL∣1,1,1-Trichloroethane 100.2 μg/mL∣Dichloroacetonitrile 100.2 μg/mL |
| Certifications/Compliance | ISO/IEC 17025 |
| Solvent or Matrix | Acetone |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?