missing translation for 'onlineSavingsMsg'
Learn More

Chlorinated Pesticides Mixture 2000 μg/mL in Toluene/Hexane, Dr. Ehrenstorfer

Product Code. 30316549
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316549 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316549 Supplier Dr. Ehrenstorfer Supplier No. DREA50000274TH

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Cis-Chlordane (Alpha Isomer)∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣Endrin-ketone∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Methoxychlor∣Trans-Chlordane (Gamma Isomer)
CAS 72-54-8, 72-55-9, 50-29-3, 309-00-2, 959-98-8, 319-84-6
Analyte or Component Names 4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Cis-Chlordane (Alpha Isomer)∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣Endrin-ketone∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Methoxychlor∣Trans-Chlordane (Gamma Isomer)
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Hexane/Toluene
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 4,4'-DDD 2000 μg/mL∣4,4'-DDE 2000 μg/mL∣4,4'-DDT 2000 μg/mL∣Aldrin 2000 μg/mL∣alpha-Endosulfan 2000 μg/mL∣alpha-HCH 2000 μg/mL∣beta-Endosulfan 2000 μg/mL∣beta-HCH 2000 μg/mL∣Cis-Chlordane (Alpha Isomer) 2000 μg/mL∣delta-HCH 2000 μg/mL∣Dieldrin 2000 μg/mL∣Endosulfan-sulfate 2000 μg/mL∣Endrin 2000 μg/mL∣Endrin-aldehyde 2000 μg/mL∣Endrin-ketone 2000 μg/mL∣gamma-HCH 2000 μg/mL∣Heptachlor 2000 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 2000 μg/mL∣Methoxychlor 2000 μg/mL∣Trans-Chlordane (Gamma Isomer) 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.