missing translation for 'onlineSavingsMsg'
Learn More

Chlorine - Cl @ 1.00 wt% in 75 cSt Mineral Oil, VHG™

Product Code. 17944036
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17944036 500 mL 500mL
17984036 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17944036 Supplier VHG Labs Supplier No. VHGCLOIL1P500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Chlorine - Cl @ 1.00 wt% in 75 cSt Mineral Oil
TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Chlorine
CAS 7782-50-5
Analyte or Component Names Chlorine
Shipping Condition Room Temperature
Molecular Formula Cl2
Concentration 1%
Certifications/Compliance ISO 17034
Synonym Cl
InChI Formula InChI=1S/Cl2/c1-2
SMILES ClCl
IUPAC Name molecular chlorine
Solvent or Matrix Heavy Mineral Oil
Molecular Weight (g/mol) 70.91
Quantity 500 mL
Formula Weight 69.9377 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.