missing translation for 'onlineSavingsMsg'
Learn More

Chlorine - Cl @ 100 μg/g in 75 cSt Mineral Oil, VHG™

Product Code. 16439649
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439649 100 mL 100mL
17911916 250 mL 250mL
17911116 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16439649 Supplier VHG Labs Supplier No. VHGCLOIL100100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Chlorine - Cl @ 100 μg/g in 75 cSt Mineral Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorine
CAS 7782-50-5
Analyte or Component Names Chlorine
Shipping Condition Room Temperature
Molecular Formula Cl2
Certifications/Compliance ISO 17034
Synonym Cl
InChI Formula InChI=1S/Cl2/c1-2
SMILES ClCl
IUPAC Name molecular chlorine
Solvent or Matrix Heavy Mineral Oil
Molecular Weight (g/mol) 70.91
Quantity 100 mL
Formula Weight 69.9377 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.