missing translation for 'onlineSavingsMsg'
Learn More

α-Chloro-m-xylene, 98%, Thermo Scientific Chemicals

Product Code. p-88305 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 10376120

Brand: Thermo Scientific undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-19-9
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000909
InChI Key LZBOHNCMCCSTJX-UHFFFAOYSA-N
Synonym 3-methylbenzyl chloride, 1-chloromethyl-3-methylbenzene, m-xylyl chloride, 3-methylbenzylchloride, alpha-chloro-m-xylene, m-methylbenzyl chloride, 1-chloromethyl-3-methyl-benzene, m-chloromethyl toluene, 3-chloromethyl toluene, benzene, 1-chloromethyl-3-methyl
PubChem CID 12102
IUPAC Name 1-(chloromethyl)-3-methylbenzene
SMILES CC1=CC(=CC=C1)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0640g/mL
Boiling Point 195.0°C to 196.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2Cl
Refractive Index 1.5335 to 1.5365
Quantity 25 mL
Beilstein 05, 373
Merck Index 15, 10289
Specific Gravity 1.064
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with absolute alcohol and ether
Formula Weight 140.61
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material α-Chloro-m-xylene, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • a-Chloro-m-xylene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.