missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene-d5, for NMR, 98.5+ atom % D

Product Code. 10398983 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10398983 1 g Glass Bottle 1g
10531241 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10398983 Supplier Thermo Scientific Acros Supplier No. 175320010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 3114-55-4 | C6H5Cl | 117.587 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3114-55-4
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 117.587
MDL Number MFCD00000531
InChI Key MVPPADPHJFYWMZ-RALIUCGRSA-N
Synonym chlorobenzene-d5, chloro 2h5 benzene, 1-chloro 2 h? benzene, chlorobenzene d5, benzene-d5, chloro, 2h5 phenyl chloride, 1-chloro 2h5 benzene, benzene-d5-, chloro, chlorobenzene, perdeutero, chlorobenzene-d5, 99 atom % d
PubChem CID 575975
IUPAC Name 1-chloro-2,3,4,5,6-pentadeuteriobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Physical Form Liquid
Color Colorless
Deuterium (D) 98.50% min.
Packaging Glass Bottle
Quantity 1 g
Linear Formula C6D5Cl
Formula Weight 117.6
Grade NMR
Chemical Name or Material Chlorobenzene-d5, 98.5+ atom % D
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorobenzene-d5
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.