missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine, 97%, maycontain suspended amine hydrochloride crystals

Product Code. 11405630
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405630 5 g 5g
11495620 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11405630 Supplier Thermo Scientific Alfa Aesar Supplier No. H25883.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 142421-57-6 | C16H6ClF12P | 492.629 g/mol

Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine is used as a reactant for the synthesis of chiral phosphine-aminophosphine ligands for rhodium-catalyzed asymmetric hydrogenation, ligands for palladium-catalyzed stereoselective allylation reactions, enantioselective hydrogenations, Josiphos analog as catalyst for asymmetric hydrogenation, ligands used in asymmetric hydrovinylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine is used as a reactant for the synthesis of chiral phosphine-aminophosphine ligands for rhodium-catalyzed asymmetric hydrogenation, ligands for palladium-catalyzed stereoselective allylation reactions, enantioselective hydrogenations, Josiphos analog as catalyst for asymmetric hydrogenation, ligands used in asymmetric hydrovinylation reactions.

Solubility
Reacts with water.

Notes
Air & moisture sensitive. Store away from oxidizing agents, bases, air, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142421-57-6
Molecular Formula C16H6ClF12P
Molecular Weight (g/mol) 492.629
MDL Number MFCD01630852
InChI Key DFZQEHBNAJGDCT-UHFFFAOYSA-N
Synonym bis 3,5-di trifluoromethyl phenyl chlorophosphine, phosphinous chloride, bis 3,5-bis trifluoromethyl phenyl, acmc-20alnu, bis 3,5-bis trifluoromethyl phenyl chlorophosphine, bis 3,5-bis trifluoromethyl phenyl-chlorophosphane, bis 3,5-bis trifluoromethyl phenyl-chlorophosphine, bis 3,5-bis trifluoromethyl phenyl chloro phosphane, p,p-bis 3,5-bis trifluoromethyl phenyl phosphinous chloride, phosphinous chloride,p,p-bis 3,5-bis trifluoromethyl phenyl
PubChem CID 5104965
IUPAC Name bis[3,5-bis(trifluoromethyl)phenyl]-chlorophosphane
SMILES C1=C(C=C(C=C1C(F)(F)F)P(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)Cl)C(F)(F)F

Specifications

Quantity 5 g
UN Number UN3265
Sensitivity Air and moisture sensitive; May form a precipitate
Solubility Information Reacts with water.
Formula Weight 492.63
Percent Purity ≥98%
Assay may contain suspended dimethylamine hydrochloride crystals
Chemical Name or Material Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.