missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis[4-(trifluoromethyl)phenyl]phosphine, 97%

Product Code. 11405640
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11405640 5 g 5g
11495630 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11405640 Supplier Thermo Scientific Alfa Aesar Supplier No. H25888.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13685-24-0 | C14H8ClF6P | 356.632 g/mol

Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13685-24-0
Molecular Formula C14H8ClF6P
Molecular Weight (g/mol) 356.632
MDL Number MFCD04972304
InChI Key PBLUOOURLNAOAJ-UHFFFAOYSA-N
Synonym bis 4-trifluoromethylphenyl chlorophosphine, chlorobis 4-trifluoromethyl phenyl phosphine, bis 4-trifluoromethyl phenyl chlorophosphine, chlorobis 4-trifluoromethyl phenyl phosphane, acmc-20alnb, chloro-bis 4-trifluoromethyl phenyl phosphane
PubChem CID 4379933
IUPAC Name chloro-bis[4-(trifluoromethyl)phenyl]phosphane
SMILES C1=CC(=CC=C1C(F)(F)F)P(C2=CC=C(C=C2)C(F)(F)F)Cl

Specifications

Boiling Point 110°C (0.4 mmHg)
Quantity 5 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 356.64
Percent Purity 97%
Chemical Name or Material Chlorobis[4-(trifluoromethyl)phenyl]phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.