missing translation for 'onlineSavingsMsg'
Learn More

Chlorodibromoacetic Acid, TRC

Product Code. 30137808
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137808 1 g 1g
30005447 100 mg 100mg
30004259 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137808 Supplier Toronto Research Chemicals Supplier No. TRCC3710381G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5278-95-5
Molecular Formula C2 H Br2 Cl O2
Molecular Weight (g/mol) 252.29
Synonym Acetic acid, 2,2-dibromo-2-chloro-, Acetic acid, dibromochloro- (7CI,8CI,9CI), 2,2-Dibromo-2-chloroacetic acid, Chlorodibromoacetic acid, Dibromochloroacetic acid
IUPAC Name 2,2-dibromo-2-chloroacetic acid
SMILES OC(=O)C(Cl)(Br)Br

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2HBr2ClO2/c3-2(4,5)1(6)7/h(H,6,7)
Formula Weight 249.8032
Chemical Name or Material Dibromochloroacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.