missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, 99.8%, for analysis, stabilized with amylene

Product Code. 10740844 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10740844 2.5 L 2.5 L
10147782 1 L 1 L
10494605 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10740844 Supplier Thermo Scientific Acros Supplier No. 383770025
  • ONLINE EXCLUSIVE PRICE
    139.00 €
    127.00 € / 2.5 L
    Save 12.00 €
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 67-66-3 | CHCl3 | 119.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3 , 513-35-9
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name chloroform
SMILES ClC(Cl)Cl

Specifications

Melting Point -63.0°C
Density 1.4800g/mL
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 5ppm max.
Assay Percent Range 99.8%
Specific Gravity 1.48
Linear Formula CHCl3
Viscosity 0.56 mPa.s (20°C)
Water 0.02% max. (K.F.)
Merck Index 15, 2142
Boiling Point 61.0°C
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone.
Formula Weight 119.38
Percent Purity 99.8%
Grade Analytical
Chemical Name or Material Chloroform, Stabilized with amylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroform, stabilized with amylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H372-Causes damage to organs.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.