missing translation for 'onlineSavingsMsg'
Learn More

Chlorophene, TRC

Product Code. 30137340
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137340 10 g 10g
30136306 25 g 25g
30003508 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137340 Supplier Toronto Research Chemicals Supplier No. TRCC37500010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-32-1
Molecular Formula C13 H11 Cl O
Molecular Weight (g/mol) 218.68
Synonym o-Cresol, 4-chloro-α-phenyl- (6CI,7CI,8CI), 4-Chloro-2-(phenylmethyl)phenol, 2-Benzyl-4-chlorophenol, 2-Hydroxy-5-chlorodiphenylmethane, 4-Chloro-2-benzylphenol, 4-Chloro-α-phenyl-o-cresol, 5-Chloro-2-hydroxydiphenylmethane, Bio-Clave, Chlorophene, Clorofene, Clorophene, Ketolin H, NSC 59989, Neosabenyl, Nipacide BCP, Nipacide BCP 50, Preventol BP, Santophen, Santophen 1, Septiphene, Septol, o-Benzyl-p-chlorophenol, p-Chloro-o-benzylphenol
IUPAC Name 2-benzyl-4-chlorophenol
SMILES Oc1ccc(Cl)cc1Cc2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H11ClO/c14-12-6-7-13(15)11(9-12)8-10-4-2-1-3-5-10/h1-7,9,15H,8H2
Formula Weight 218.0498
Chemical Name or Material 2-Benzyl-4-chlorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.