missing translation for 'onlineSavingsMsg'
Learn More

Chloroquine phosphate, MedChemExpress

Product Code. 16447386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16447386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16447386 Supplier Medchem Express Supplier No. HY17589
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Chloroquine phosphate is an antimalarial and anti-inflammatory agent widely used to treat malaria and rheumatoid arthritis. Chloroquine phosphate is an autophagy and toll-like receptors (TLRs) inhibitor. Chloroquine phosphate is highly effective in the control of SARS-CoV-2 (COVID-19) infection in vitro (EC50=1.13 μM).

Chloroquine phosphate is an antimalarial and anti-inflammatory agent widely used to treat malaria and rheumatoid arthritis. Chloroquine phosphate is an autophagy and toll-like receptors (TLRs) inhibitor. Chloroquine phosphate is highly effective in the control of SARS-CoV-2 (COVID-19) infection in vitro (EC50=1.13 μM).

Specifications

Chemical Name or Material Chloroquine phosphate
Physical Form Powder
CAS 50-63-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H32ClN3O8P2
For Use With (Application) COVID-19-anti-virus
Solubility Information H2O : ≥ 33 mg/mL (63.97 mM) ∣DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble)
SMILES O=P(O)(O)O.O=P(O)(O)O.CC(NC1=CC=NC2=CC(Cl)=CC=C12)CCCN(CC)CC
Molecular Weight (g/mol) 515.86
Formula Weight 515.86
Percent Purity 99.31%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.